1,2-Epoxycyclopentane 98%

Supplier: Thermo Scientific Chemicals
Danger

34852-0250 34852-1000
TS34852-0250EA 313 USD
TS34852-0250 TS34852-1000
1,2-Epoxycyclopentane 98%
1,2-Epoxycyclopentane
Formula: C₅H₈O
Boiling Pt: 102 °C (760 mmHg)
Density: 0.964 g/cm³
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005161
CAS Number: 285-67-6
EINECS: 206-005-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4360 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR