Harmine 98%

Supplier: Thermo Scientific Chemicals
Warning

30297-0010 30297-0050 30297-2500
TS30297-0010EA 125 USD
TS30297-0010 TS30297-0050 TS30297-2500
Harmine 98%
Harmine
Formula: C₁₃H₁₂N₂O
Melting Pt: 262…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00004958
CAS Number: 442-51-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR