2-Phenylquinoline 99+%

Supplier: Thermo Scientific Chemicals
Warning

29799-0010 29799-0050
TS29799-0010EA 77 USD
TS29799-0010 TS29799-0050
2-Phenylquinoline 99+%
2-Phenylquinoline
Formula: C₁₅H₁₁N
Boiling Pt: 363 °C (760 mmHg)
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00011568
CAS Number: 612-96-4
EINECS: 210-326-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 82 to 84 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR