2-Amino-1,3,5-triazine 98%

Supplier: Thermo Scientific Chemicals
Warning

29524-0010 29524-0050
TS29524-0010EA 83 USD
TS29524-0010 TS29524-0050
2-Amino-1,3,5-triazine 98%
2-Amino-1,3,5-triazine
Formula: C₃H₄N₄
Melting Pt: 219…224 °C
MDL Number: MFCD00023179
CAS Number: 4122-04-7
EINECS: 223-920-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR