meso-Tetraphenylporphine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP
17121-0010
17121-0050
17121-0250
TS17121-0010EA
67
USD
TS17121-0010
TS17121-0050
TS17121-0250
meso-Tetraphenylporphine 97%
meso-Tetraphenylporphine
Formula:
C₄₄H₃₀N₄ MW: 614.75 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011680 CAS Number: 917-23-7 EINECS: 213-025-9 |
Specification Test Results
Appearance | Blue to purple Crystalline powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Impurity | ≤3 % Chlorin |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...