3-(2-Hydroxyethyl)indole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol

14060-0010 14060-0050
TS14060-0010EA 29 USD
TS14060-0010 TS14060-0050
3-(2-Hydroxyethyl)indole 97%
3-(2-Hydroxyethyl)indole
Formula: C₁₀H₁₁NO
Boiling Pt: 174 °C (2.7mbar)
Melting Pt: 57…60 °C
Flash Pt: 255 °C
MDL Number: MFCD00005659
CAS Number: 526-55-6
EINECS: 208-393-2
Merck Index: 12,09930

Order Now

Specification Test Results

Appearance Off-white to brown Powder or crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 56 to 61 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR