Melamine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
12535-0010
12535-0050
12535-0100
12535-1000
TS12535-0010EA
44
USD
TS12535-0010
TS12535-0050
TS12535-0100
TS12535-1000
Melamine 99%
Melamine
Formula:
C₃H₆N₆ MW: 126.12 g/mol Melting Pt: 354 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006055 CAS Number: 108-78-1 EINECS: 203-615-4 Merck Index: 13,05834 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...