Chromone-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid
12388-0050
12388-0250
TS12388-0050EA
37
USD
TS12388-0050
TS12388-0250
Chromone-2-carboxylic acid 97%
Chromone-2-carboxylic Acid
Formula:
C₁₀H₆O₄ Melting Pt: 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006838 CAS Number: 4940-39-0 EINECS: 225-583-0 |
Specification Test Results
Appearance | White to beige to grey-brown Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...