(±)-3-Hydroxytetrahydrofuran 97%

Supplier: Thermo Scientific Chemicals
Warning

12194-0050 12194-0250 12194-1000
TS12194-0050EA 39 USD
TS12194-0050 TS12194-0250 TS12194-1000
(±)-3-Hydroxytetrahydrofuran 97%
(±)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
Boiling Pt: 181 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005374
CAS Number: 453-20-3
EINECS: 207-219-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥96.0 %
UV at 240 nm A: ≤0.05
Titration after Ion exchange ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR