2-Hydroxypyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone

12184-0050 12184-0500 12184-2500
TS12184-0050EA 23 USD
TS12184-0050 TS12184-0500 TS12184-2500
2-Hydroxypyridine 97%
2-Hydroxypyridine
Formula: C₅H₅NO
Boiling Pt: 280…281 °C (760 mmHg)
Melting Pt: 102…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00006268
CAS Number: 142-08-5
EINECS: 205-520-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to yellow to beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 103 to 108 °C
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR