Flavone 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Phenylchromen-4-one

11916-0010 11916-0050 11916-0250
TS11916-0010EA 29 USD
TS11916-0010 TS11916-0050 TS11916-0250
Flavone 99+%
Flavone
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Melting Pt: 96
Storage Temperature: Ambient
MDL Number: MFCD00006825
CAS Number: 525-82-6
EINECS: 208-383-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 95 to 98 °C
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR