2,5-Dimercapto-1,3,4-thiadiazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3,4-Thiadiazole-2,5-dithiol
11531-0050
11531-0250
11531-1000
TS11531-0050EA
22
USD
TS11531-0050
TS11531-0250
TS11531-1000
2,5-Dimercapto-1,3,4-thiadiazole 98%
2,5-Dimercapto-1,3,4-thiadiazole
Formula:
C₂H₂N₂S₃ MW: 150.25 g/mol Melting Pt: 161…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003103 CAS Number: 1072-71-5 EINECS: 214-014-1 |
Specification Test Results
Appearance | Off-white to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 156 to 162 °C (Dec) |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...