1,2-Epoxycyclohexane 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Epoxycyclohexane, Cyclohexene oxide

11121-1000 11121-5000
TS11121-1000EA 24 USD
TS11121-1000 TS11121-5000
1,2-Epoxycyclohexane 98%
1,2-Epoxycyclohexane
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129
Melting Pt: -27
Density: 0.97 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR