5-Bromo-2-furoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid

10702-0250 10702-1000
TS10702-0250EA 47 USD
TS10702-0250 TS10702-1000
5-Bromo-2-furoic acid 99%
5-Bromo-2-furoic acid
Formula: C₅H₃BrO₃
MW: 190.98 g/mol
Melting Pt: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00003239
CAS Number: 585-70-6

Order Now

Specification Test Results

Appearance White to cream to beige to brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 188 to 192 °C
Titration with NaOH ≥98.5 %
Color scale ≤10 APHA
Stabilizer 5 to 30 ppm BHT
Residue after evaporation ≤0.005 %
Carbonyl (as HCHO) ≤0.01 %
Titrable acid ≤0.0016 meq/g
Peroxides ≤0.005 % (H2O2)
Freezing point ≥11.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR