Black Diamond Digital Heating Mantles With Stirrer, United Scientific Supplies
Supplier: United Scientific Supplies
UNDHMTLS-100 UNDHMTLS-1000 UNDHMTLS-2000 UNDHMTLS-250 UNDHMTLS-3000 UNDHMTLS-500 UNDHMTLS-5000
470339-110EA 788 USD
470339-110 470339-576 470339-630 470339-604 470339-980 470339-582 470339-918
Black Diamond Digital Heating Mantles With Stirrer, United Scientific Supplies
Heaters Heating Mantles
Digital control system for heating and stirring.
  • Stainless steel and aluminum construction
  • Separate control knobs for heating and stirring
  • Digital display
  • External probe to control the flask temperature
  • Max. temperature: 400 °C
This series of digital heating mantles includes a built-in stirrer function. Separate knobs control the temperature level up to 400 °C and stirring speed up to 800 rpm. The external probe allows the temperature of the contents of the flask to be directly measured. Case is constructed of durable stainless steel and aluminum materials. Knitted mantle is made from glass fiber yarn and designed to securely hold round bottom flasks with the top half of the flask visible during operation. Available in various sizes to hold flasks from 100 to 5000 ml capacity. Includes a spill collect system to drain solution if a flask breaks during use.