(±)-1,2-Propanediol ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
44741-0025
44741-1000
44741-5000
TS44741-0025EA
241
USD
TS44741-0025
TS44741-1000
TS44741-5000
(±)-1,2-Propanediol ACS
Propylene glycol
Formula:
CH₃CH(OH)CH₂OH MW: 76.1 g/mol Boiling Pt: 187.6 Melting Pt: -59 Density: 1.03 g/cm³ Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 Merck Index: 13,07947 |
|
Specification Test Results
Appearance | Clear viscous liquid |
Water | ≤0.2 % |
GC | ≥99.5 % |
Color scale | ≤10 APHA |
Residue after ignition | ≤0.005 % |
Chloride (Cl) | ≤1 ppm |
Titrable acid | ≤0.0005 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...