Isoamyl alcohol ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol
41273-0010
41273-0025
41273-5000
TS41273-0010EA
151
USD
TS41273-0010
TS41273-0025
TS41273-5000
Isoamyl alcohol ACS
Isoamyl alcohol
Formula:
(CH₃)₂CHCH₂CH₂OH MW: 88.15 g/mol Boiling Pt: 130 Melting Pt: -117 Density: 0.8 g/cm³ Flash Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3 III Merck Index: 13,05214 |
|
Specification Test Results
Appearance | Clear liquid |
Water | ≤0.5 % |
GC | ≥98.5 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.003 % |
Carbonyl (as HCHO) | ≤0.1 % |
Acids and esters | ≤0.2 % (as Amyl acetate) |
Titrable acid | ≤0.002 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...