p-Hydrazinobenzenesulfonic acid hemihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydrazinobenzenesulphonic acid, hemihydratePhenylhydrazine-4-sulfonic acid4-Hydrazinobenzenesulfonic acid, hemihydratePhenylhydrazine-4-sulphonic acid

41178-0500 41178-1000
TS41178-1000EA 149 USD
TS41178-1000 TS41178-0500
p-Hydrazinobenzenesulfonic acid hemihydrate 98%
p-Hydrazinobenzenesulfonic acid hemihydrate
Formula: C₆H₈N₂O₃S
MW: 188.20 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00480958
CAS Number: 854689-07-9

Order Now

Specification Test Results

Appearance Light yellow to light orange Powder
Infrared spectrum Conforms
Loss on drying 4 % to 5 %
Titration with NaOH ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR