4-Hydroxythiophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

37063-0050 37063-0250
TS37063-0050EA 61 USD
TS37063-0050 TS37063-0250
4-Hydroxythiophenol 99%
4-Hydroxythiophenol
Formula: C₆H₆OS
Boiling Pt: 149…150 °C (25 mmHg)
Melting Pt: 33…35 °C
Flash Pt: 110 °C
MDL Number: MFCD00004850
CAS Number: 637-89-8
EINECS: 211-307-6
UN: 2923

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR