4-Fluorobenzyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

31930-0050 31930-0010
TS31930-0050EA 358 USD
TS31930-0050 TS31930-0010
4-Fluorobenzyl isocyanate 98%
4-Fluorobenzyl isocyanate
Formula: C₈H₆FNO
Boiling Pt: 211 °C (760 mmHg)
Density: 1.186 g/cm³
Flash Pt: 93 °C
Storage Temperature: Freezer
MDL Number: MFCD00673062
CAS Number: 132740-43-3
UN: 2206
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5060 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR