4-Chlorophenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
16656-0250
16656-1000
16656-5000
TS16656-0250EA
39
USD
TS16656-0250
TS16656-1000
TS16656-5000
4-Chlorophenyl isocyanate 98%
4-Chlorophenyl isocyanate
Formula:
C₇H₄ClNO MW: 153.57 Boiling Pt: 203 Melting Pt: 29…31 °C Density: 1.26 g/cm³ Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002024 CAS Number: 104-12-1 EINECS: 203-176-9 UN: 2811 ADR: 6.1 II |
Specification Test Results
Appearance | White to yellow Crystalline solid (May develop some turbidity or precipitate) |
Infrared spectrum | Conforms |
Melting point | 29 to 31 °C |
GC | ≥97.5 % |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...