Pyrocatechol 99+%

Supplier: Thermo Scientific Chemicals
Danger

15898-0025 15898-0050 15898-0100 15898-1000 15898-5000
TS15898-0025EA 214 USD
TS15898-0025 TS15898-0050 TS15898-0100 TS15898-1000 TS15898-5000
Pyrocatechol 99+%
Pyrocatechol
Formula: C₆H₆O₂
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: 103…106 °C
Flash Pt: 131 °C
Storage Temperature: Ambient
MDL Number: MFCD00002188
CAS Number: 120-80-9
EINECS: 204-427-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,08089

Order Now

Specification Test Results

Appearance Off-white to grey-brown Crystalline flakes (May darken on storage)
Infrared spectrum Conforms
Melting point 103 to 106 °C
GC ≥99.0 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.05 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR