(±)-2-Methyl-2,4-pentanediol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexylene glycol, (±)-2,4-Dihydroxy-2-methyl pentane, (±)-2,4-Dihydroxy-2-methyl pentane, 2-Methylpentane-2,4-diol, Hexylene Glycol

15034-0010 15034-0025 15034-0250
TS15034-0010EA 45 USD
TS15034-0010 TS15034-0025 TS15034-0250
(±)-2-Methyl-2,4-pentanediol 99%
(±)-2-Methyl-2,4-pentanediol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 198
Melting Pt: -40
Density: 0.922 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS Number: 107-41-5
EINECS: 203-489-0
Merck Index: 13,04730

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤15 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR