α,4-Dichlorotoluene 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

15024-0010 15024-0050 15024-2500
TS15024-0010EA 158 USD
TS15024-0010 TS15024-0050 TS15024-2500
α,4-Dichlorotoluene 99+%
α,4-Dichlorotoluene
Formula: C₇H₆Cl₂
Boiling Pt: 221 °C (760 mmHg)
Melting Pt: 27…28 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to white Low melting crystalline solid
Infrared spectrum Conforms
Melting point ≥28 °C
GC ≤0.5 % p-Chlorobenzal chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR