2,4-Dichlorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15022-0050
15022-1000
15022-5000
TS15022-0050EA
28
USD
TS15022-0050
TS15022-1000
TS15022-5000
2,4-Dichlorobenzoyl chloride 98%
2,4-Dichlorobenzoyl chloride
Formula:
C₇H₃Cl₃O Boiling Pt: 150 °C (34 mmHg) Melting Pt: 21 °C Density: 1.49 g/cm³ Flash Pt: 137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000661 CAS Number: 89-75-8 EINECS: 201-936-4 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to slightly colored Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5915 to 1.5935 (20°C, 589 nm) |
GC | ≥97.5 % |
Titration RCOCl | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...