4-Methoxythiophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

14915-0050 14915-0250
TS14915-0050EA 68 USD
TS14915-0050 TS14915-0250
4-Methoxythiophenol 98%
4-Methoxythiophenol
Formula: C₇H₈OS
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.14 g/cm³
Flash Pt: 96 °C
MDL Number: MFCD00004849
CAS Number: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5850 (20°C, 589 nm)
GC ≥97.5 %
TLC ≥95 %
Appearance of solution (0.5mmol/ml DMF) clear, colourless
Specific optical rotation +9.0° to +11.0° (20°C, 589 nm) (c=1 in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR