Cinnamyl alcohol ≥98% (trans-isomers)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol

14852-0050 14852-1000 14852-2500 14852-5000
TS14852-0050EA 30 USD
TS14852-0050 TS14852-1000 TS14852-2500 TS14852-5000
Cinnamyl alcohol ≥98% (trans-isomers)
Cinnamyl alcohol
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 256
Melting Pt: 31…35 °C
Density: 1.04 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325

Order Now

Specification Test Results

Appearance White to light yellow Fused crystalline solid
Infrared spectrum Conforms
Melting point 31 to 35 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR