Rubrene 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

13238-0010 13238-5000
TS13238-0010EA 162 USD
TS13238-0010 TS13238-5000
Rubrene 99%
Rubrene
Formula: C₄₂H₂₈
MW: 532.68 g/mol
Melting Pt: 330…335 °C
MDL Number: MFCD00003703
CAS Number: 517-51-1
EINECS: 208-242-0

Order Now

Specification Test Results

Appearance Red Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR