Pentafluorobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

12991-0050 12991-0250 12991-1000
TS12991-0050EA 29 USD
TS12991-0050 TS12991-0250 TS12991-1000
Pentafluorobenzoic acid 99%
Pentafluorobenzoic acid
Formula: C₇HF₅O₂
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002406
CAS Number: 602-94-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 104 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR