Pentacene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,3,6,7-Dibenzanthracene

12979-0010 12979-1000
TS12979-0010EA 286 USD
TS12979-0010 TS12979-1000
Pentacene 98%
Pentacene
Formula: C₂₂H₁₄
MW: 278.35 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003710
CAS Number: 135-48-8
EINECS: 205-193-7
Merck Index: 12,07240

Order Now

Specification Test Results

Appearance Dark blue to purple to black Crystalline powder
Infrared spectrum Conforms
Assay ≥97.5 %
Particle size ≤4 % (< 40 mesh)
Molasses number ≥230
Abrasion number ≥75
Iodine number ≥1000 mg/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR