1,8-Octanediol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,8-Dihydroxy octane
12938-0250
12938-1000
TS12938-0250EA
41
USD
TS12938-0250
TS12938-1000
1,8-Octanediol 98%
1,8-Octanediol
Formula:
C₈H₁₈O₂ Boiling Pt: 278.8 °C (760 mmHg) Melting Pt: 60…62 °C Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002989 CAS Number: 629-41-4 EINECS: 211-090-8 |
Specification Test Results
Appearance | Colorless to white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 58 to 63 °C |
Water | ≤0.5 % (K.F.) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...