2-Fluorobenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
o-Fluorobenzoic acid
11936-0250
11936-1000
TS11936-0250EA
39
USD
TS11936-0250
TS11936-1000
2-Fluorobenzoic acid 99%
2-Fluorobenzoic acid
Formula:
C₇H₅FO₂ Boiling Pt: 245 °C (1013 hPa) Melting Pt: 123…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 EINECS: 207-158-1 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 122 to 125 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...