Ethyl benzoate 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzoic acid ethyl ester
11809-0025
11809-5000
TS11809-0025EA
113
USD
TS11809-0025
TS11809-5000
Ethyl benzoate 99+%
Ethyl benzoate
Formula:
C₉H₁₀O₂ Boiling Pt: 415 °C (1013 hPa) Melting Pt: –34 °C Density: 1.045 g/cm³ Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009109 CAS Number: 93-89-0 EINECS: 202-284-3 Merck Index: 13,03802 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5040 to 1.5060 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific gravity | (25/25°C) 1.043 to 1.047 |
Acid value | ≤1 mg KOH/g |
Chlorate and Nitrate (as NO3) | ≤0.003 % |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤0.004 % |
Barium (Ba) | Passes test |
Calcium (Ca) | ≤0.002 % |
Iron (Fe) | ≤2 ppm |
Magnesium (Mg) | ≤0.001 % |
Potassium (K) | ≤0.005 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...