Diphenyl chlorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diphenyl phosphorochloridate

11724-0250 11724-1000 11724-5000
TS11724-0250EA 32 USD
TS11724-0250 TS11724-1000 TS11724-5000
Diphenyl chlorophosphate 98%
Diphenyl chlorophosphate
Formula: C₁₂H₁₀ClO₃P
MW: 268.64 g/mol
Boiling Pt: 314…316 °C (272 mmHg)
Density: 1.29 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003030
CAS Number: 2524-64-3
EINECS: 219-759-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5485 to 1.5505 (20°C, 589 nm)
GC ≥97.5 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g
Thiophene Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR