Coronene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hexabenzobenzene
11051-1000
11051-5000
TS11051-1000EA
84
USD
TS11051-1000
TS11051-5000
Coronene 95%
Coronene
Formula:
C₂₄H₁₂ MW: 300.36 g/mol Boiling Pt: 525 Melting Pt: 438…440 °C |
MDL Number:
MFCD00004134 CAS Number: 191-07-1 EINECS: 205-881-7 UN: 2811 |
Specification Test Results
Appearance | Yellow to green Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Titration RCOCl | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...