4-tert-Butylphenol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10800-0010
10800-0025
10800-0050
TS10800-0010EA
69
USD
TS10800-0010
TS10800-0025
TS10800-0050
4-tert-Butylphenol 97%
4-tert-Butylphenol
Formula:
C₁₀H₁₄O MW: 150.22 g/mol Boiling Pt: 237 Melting Pt: 96…100 °C Density: 0.9 g/cm³ Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002367 CAS Number: 98-54-4 EINECS: 202-679-0 UN: 3077 ADR: 9 III Merck Index: 13,01584 |
Specification Test Results
Appearance | White to light beige Flakes or pastilles |
Infrared spectrum | Conforms |
Melting point | 96 to 100 °C |
Water | ≤0.5 % (K.F.) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...