Benzoic anhydride 98%

Supplier: Thermo Scientific Chemicals
Danger

10550-0025 10550-0250 10550-1000 10550-5000
TS10550-0025EA 467 USD
TS10550-0025 TS10550-0250 TS10550-1000 TS10550-5000
Benzoic anhydride 98%
Benzoic anhydride
Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Boiling Pt: 360
Melting Pt: 40
Density: 1.19 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003073
CAS Number: 93-97-0
EINECS: 202-291-1
Merck Index: 13,01093

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 38 to 44 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR