Benzhydryl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10539-1000
10539-5000
TS10539-1000EA
47
USD
TS10539-1000
TS10539-5000
Benzhydryl alcohol 99%
Benzhydryl alcohol
Formula:
C₁₃H₁₂O Boiling Pt: 297 °C (1013 hPa) Melting Pt: 65…68 °C Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004488 CAS Number: 91-01-0 EINECS: 202-033-8 Merck Index: 12,01121 |
Specification Test Results
Appearance | White to beige Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 65 to 68 °C |
Water | ≤0.5 % (K.F.) |
Loss on drying | ≤0.5 % (55 °C) |
GC | ≥98.5 % |
Lead (Pb) | ≤10 ppm |
Nickel (Ni) | ≤100 ppm |
Potassium (K) | ≤50 ppm |
Sodium (Na) | ≤100 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...