4-Allylanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Allyl-4-methoxybenzene, Estragole

10288-0050 10288-0250 10288-1000
TS10288-0050EA 18 USD
TS10288-0050 TS10288-0250 TS10288-1000
4-Allylanisole 98%
4-Allylanisole
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 22.5 °C
Density: 0.96 g/cm³
Flash Pt: 81 °C
MDL Number: MFCD00008653
CAS Number: 140-67-0
EINECS: 205-427-8
Merck Index: 13,03740

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5240 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +20.5° to +21.5° (25°C, 589 nm) (c=10, H2O)
Iron (Fe) ≤10 ppm
Heavy metals (ICP-OES) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR