United Scientific™ Spectrum Tube
Supplier: United Scientific Supplies
SPTNI01 SPTOX01 SPTWV01 SPTXE01 SPTAI01 SPTAR01 SPTCD01 SPTHE01 SPTHY01 SPTKR01 SPTNE01
470338-962EA 44.6 USD
470338-962 470340-256 470339-546 470339-810 470339-252 470339-520 470339-338 470339-326 470338-954 470339-824 470340-088 470339-328
United Scientific™ Spectrum Tube
Educational Materials Physics Educational Materials Optics Learning Activities
Investigate the spectral pattern of different gases.
  • Ideal for classroom demonstrations of spectral lines
  • Glass tube is 25 cm long, with 10 cm long capillary portion
  • Electrode sealed in each end, attached to metal end cap with loop for convenient hookup to wire connections
Air spectrum tube for classroom demonstrations of spectral lines. Spectrum tubes acquaint students with the spectra of common gases in their pure form, and are also used for demonstrating wavelength. Each gas has its own distinctive set of spectral lines when energized and viewed through a spectroscope.
Glass tube measures 25 cm long with the capillary portion about 5 cm long. An electrode is sealed in each end and attached to a metal end cap with a loop for convenient hookup to wire connections.
Ordering information: A spectrum tube power supply (not included) is required to energize the tube.
Delivery information: Includes an activity guide.