Innovating Science® Diffusion and Cell Size Lab Activity
Supplier: Ward's Science
IS3003-SGL IS3003
470336-180EA 22.5 USD
470336-180 470343-472
Innovating Science® Diffusion and Cell Size Lab Activity
Educational Classroom Kits and Activities
Learn why cells are microscopic and the need for cells to effectively move materials in and remove waste.
  • Students whill create simulated cells of different sizes and examine how effectively a substance is able to diffuse into the cell
  • Aligned to NGSS Practices
  • Performance expectations: MS-LS1-1, MS-LS1-2
  • Disciplinary core ideas: LS1.A
  • Cross cutting concepts include scale proportion and quantity, structure and function
  • Engineering practices include developing and using models, planning and carrying out investigations
Why are cells microscopic? The answer relates to the needs for the cell to effectively move materials in and remove waste. In this activity, students will create simulated cells (agar blocks) of different sizes and examine how effectively a substance is able to diffuse into the cell in a set period of time. A special indicator in the cells will allow students to visualize the degree of diffusion.
The results will clearly display the fact that a smaller volume creates a more favorable condition for the exchange of material across a cell membrane. Kit contains enough materials for 15 groups of students. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Instructor's Manual and SDS included.