Calcium stearate (6,4 - 7,4% Ca), powder

Supplier: Thermo Scientific Chemicals

Synonyms: Stearic acid calcium salt

44972-0010 44972-0050
TS44972-0010EA 61 USD
TS44972-0010 TS44972-0050
Calcium stearate (6,4 - 7,4% Ca), powder
Calcium stearate
Formula: C₃₆H₇₀CaO₄
Melting Pt: 145…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00036390
CAS Number: 1592-23-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration Complexometric 6.4 to 7.5 % (Ca)
Loss on drying ≤6 % (105°C, 3 hrs)
Ash 9 % to 10.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR