Lithium stearate 95%

Supplier: Thermo Scientific Chemicals

41334-0250 41334-1000
TS41334-0250EA 31 USD
TS41334-0250 TS41334-1000
Lithium stearate 95%
Lithium stearate
Formula: C₁₈H₃₅LiO₂
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00042032
CAS Number: 4485-12-5
EINECS: 224-772-5

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR