4-Methylumbelliferyl-β-D-galactopyranoside

Supplier: Thermo Scientific Chemicals

33721-0010 33721-0050
TS33721-0010EA 133 USD
TS33721-0010 TS33721-0050
4-Methylumbelliferyl-β-D-galactopyranoside
4-Methylumbelliferyl-β-D-galactopyranoside
Formula: C₁₆H₁₈O₈
Melting Pt: 248 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036773
CAS Number: 6160-78-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
TLC Complies
Appearance of solution (1% in pyridine) clear colorless
Specific optical rotation -62° to -66° (20°C, 589 nm) (c=1, pyridine, after 24h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR