4-Hydroxy-2-methoxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
33593-0010
33593-0050
33593-0250
TS33593-0010EA
60
USD
TS33593-0010
TS33593-0050
TS33593-0250
4-Hydroxy-2-methoxybenzaldehyde 98%
4-Hydroxy-2-methoxybenzaldehyde
Formula:
C₈H₈O₃ Melting Pt: 153…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051964 CAS Number: 18278-34-7 |
Specification Test Results
Appearance | Off-white to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 156 to 161 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...