2,6-Diacetylpyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

33086-0010 33086-0250 33086-0050
TS33086-0010EA 49 USD
TS33086-0010 TS33086-0250 TS33086-0050
2,6-Diacetylpyridine 99%
2,6-Diacetylpyridine
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00006304
CAS Number: 1129-30-2
EINECS: 214-442-9

Order Now

Specification Test Results

Appearance White to beige Powder or crystals or needles
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR