Cyclopropylméthanol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol

29682-0010 29682-0250 29682-1000 29682-2500
TS29682-0010EA 752 USD
TS29682-0010 TS29682-0250 TS29682-1000 TS29682-2500
Cyclopropylméthanol 99%
Cyclopropylméthanol
Formula: C₄H₈O
Boiling Pt: 123…124 °C (738 mmHg)
Melting Pt: –60 °C
Density: 0.89 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001309
CAS Number: 2516-33-8
EINECS: 219-735-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR