2-Furaldehyde 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Furfurol, Furfural

18110-0010 18110-0025 18110-0100 18110-0250 18110-2500
TS18110-0010EA 114 USD
TS18110-0010 TS18110-0025 TS18110-0100 TS18110-0250 TS18110-2500
2-Furaldehyde 99%
2-Furaldehyde
Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 162
Melting Pt: -39
Density: 1.16 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1 II
Merck Index: 13,04325

Order Now

Specification Test Results

Appearance Clear amber to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Residue after evaporation ≤0.5 %
Acidity ≤0.02 eq/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR