6-Bromocaproic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Bromohexanoic acid

16567-0250 16567-1000 16567-5000
TS16567-0250EA 74 USD
TS16567-0250 TS16567-1000 TS16567-5000
6-Bromocaproic acid 98%
6-Bromocaproic acid
Formula: BrH₂C(CH₂)₄COOH
MW: 195.06 g/mol
Boiling Pt: 165
Melting Pt: 32…35 °C
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light orange Low melting crystals
Infrared spectrum Conforms
Melting point 32 to 35 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR