Indole-3-carbaldehyd 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde
15664-0050
15664-0250
15664-1000
TS15664-0050EA
31
USD
TS15664-0050
TS15664-0250
TS15664-1000
Indole-3-carbaldehyd 97%
Indole-3-carbaldehyd
Formula:
C₉H₇NO MW: 145.16 g/mol Melting Pt: 193…199 °C Flash Pt: 240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005622 CAS Number: 487-89-8 EINECS: 207-665-8 |
Specification Test Results
Appearance | Off-white to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 193 to 199 °C |
HPLC | ≥96.0 % |
UV | at 500 nm A: ≤0.01 |
pH | 5.0 to 9.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...